![]() |
The Talos™ L120C is a new high-contrast, 120kV thermionic , multi-discipline transmission/scanning transmission electron microscope (S/TEM) designed for both life and materials scientists. Incorporating proven technologies from Thermo Fisher Scientific's high-end Titan Krios™/Themis™ S/TEM family, the Talos L120C improves upon its predecessor, the Tecnai™ Spirit, by improving performance and usability in laboratories with users of varying skill levels.
For Life Sciences, the Talos L120C provides improved performance and productivity across a wide range of samples and applications, such as 2D and 3D imaging of cells and cell organelles, at both ambient and cryogenic temperatures.
Why should I watch this webcast?